Learn More
Epredia™ Cassette II Slotted Tissue Cassettes
Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Beskrivning
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifikationer
Specifikationer
| Pore Style | Slots |
| Closure Included | Yes |
| Quantity | 1000/Cs. |
| Closure Type | Separate Lid, Front Hinge |
| Color | Lilac |
| Product Type | Slotted Cassette |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering