Learn More
Tocris Bioscience™ CASIN
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
CASIN is a Cdc42 GTPase inhibitor (IC50 = 2 μM). Reduces active levels of Cdc42 in aged hematopoietic stem cells (HSCs) to that in young HSCs. Reverses the aging-related and polarity phenotype of aged HSCs to that of young HSCs in vivo. Increases histone H4K16 acetylation in aged HSCs.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | CASIN |
| Purity | >98% |
| Molecular Weight (g/mol) | 306.4 |
| Inhibitors | Rho GTPase Inhibitors |