Learn More
Carvacrol, TRC
Product Code. 30005233
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005233 5 g 5g
30012463 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30005233 Supplier Toronto Research Chemicals Supplier No. TRCC1846005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 499-75-2
Molecular Formula C10 H14 O
Molecular Weight (g/mol) 150.22
Synonym Phenol, 2-methyl-5-(1-methylethyl)-, Carvacrol (8CI), 2-Hydroxy-1-methyl-4-(1-methylethyl)benzene, 2-Hydroxy-p-cymene, 2-Methyl-5-(1-methylethyl)phenol, 2-Methyl-5-(propan-2-yl)phenol, 2-Methyl-5-isopropylphenol, 3-Isopropyl-6-methylphenol, 5-Isopropyl-2-methylphenol, 5-Isopropyl-o-cresol, 6-Methyl-3-isopropylphenol, Antioxine, Dentol, Isopropyl o-cresol, Isothymol, NSC 6188, p-Cymen-2-ol
IUPAC Name 2-methyl-5-propan-2-ylphenol
SMILES CC(C)c1ccc(C)c(O)c1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4-7,11H,1-3H3
Formula Weight 150.1045
Chemical Name or Material Carvacrol