Learn More
Hanna Instruments™ Carrying Case
Product Code. 11584457
Change view
Click to view available options
Dimensions (L x W x H):
340 x 230 x 90 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11584457 340 x 230 x 90 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11584457 Supplier Hanna Instruments™ Supplier No. HI710031
  • ONLINE EXCLUSIVE PRICE
    128.60€
    77.85€ / Pack of 1
    Save 50.75€
    39% Off
Estimated Shipment: 15-12-2026
Specifications
Product Type Carry Case
Dimensions (L x W x H) 340 x 230 x 90 mm
For Use With (Equipment) For use with HI 93510 meter