Learn More
Carphenazine Dimaleate, TRC
Specifications
Specifications
| Chemical Name or Material | Carpenazine Dimaleate |
| CAS | 2975-34-0 |
| Molecular Formula | C24 H31 N3 O2 S . 2 C4 H4 O4 |
| Quantity | 5 mg |
| Synonym | 1-Propanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-, (2Z)-2-butenedioate (1:2), 1-Propanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-, (2Z)-2-butenedioate (1:2) (salt) (9CI), 1-Propanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-, (Z)-2-butenedioate (1:2), 1-Propanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl]-, dimaleate (6CI), 1-Propanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl]-, maleate (1:2) (salt) (8CI), 1-Propanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl]-, maleate (7CI), 1-(10-(3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propyl)phenothiazin-2-yl)-1-propanone dimaleate, 1-[10-[3-[4-(2-Hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl]-1-propanone bis(hydrogen maleate), Carfenazine maleate, Carphenazine maleate, NSC 71755, Proketazine maleate, Wy 2445 |
| InChI Formula | InChI=1S/C24H31N3O2S.2C4H4O4/c1-2-22(29)19-8-9-24-21(18-19)27(20-6-3-4-7-23(20)30-24)11-5-10-25-12-14-26(15-13-25)16-17-28;2*5-3(6)1-2-4(7)8/h3-4,6-9,18,28H,2,5,10-17H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1- |
| SMILES | CCC(=O)c1ccc2Sc3ccccc3N(CCCN4CCN(CCO)CC4)c2c1.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;1-[10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazin-2-yl]propan-1-one |
| Molecular Weight (g/mol) | 657.73 |
| Formula Weight | 657.2356 |