missing translation for 'onlineSavingsMsg'
Learn More

Carnauba Wax, TRC

Product Code. 30136219
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136219 250 g 250g
30010744 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136219 Supplier Toronto Research Chemicals Supplier No. TRCC184085250G
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8015-86-9
Synonym Carnauba wax substitutes (03C, 04C, 05C, 06C), Waxes, carnauba (01C, 02C), Carnauba wax (03C, 04C, 05C, 06C, 07C, 08C, 09C, 10C, 11C, 14C, 15C), Copernicia cerifera wax

Specifications

Quantity 250 g
Chemical Name or Material Carnauba wax
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.