Learn More
Carmoterol Hydrochloride, TRC
Product Code. 30180072
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180072 1 mg 1mg
30180115 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30180072 Supplier Toronto Research Chemicals Supplier No. TRCC1837501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 137888-11-0
Molecular Formula C21 H24 N2 O4 . Cl H
Molecular Weight (g/mol) 404.89
Synonym 2(1H)-Quinolinone, 8-hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]-, hydrochloride (1:1), 2(1H)-Quinolinone, 8-hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]-, monohydrochloride (9CI), 2(1H)-Quinolinone, 8-hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]-, monohydrochloride, [R-(R*,R*)]-, (R,R)-Carmoterol hydrochloride, CHF 4226.01, Carmoterol hydrochloride, TA 2005, 8-Hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]-2(1H)-quinolinone hydrochloride
IUPAC Name 8-hydroxy-5-[(1R)-1-hydroxy-2-[[(2R)-1-(4-methoxyphenyl)propan-2-yl]amino]ethyl]-1H-quinolin-2-one;hydrochloride
SMILES Cl.COc1ccc(C[C@@H](C)NC[C@H](O)c2ccc(O)c3NC(=O)C=Cc23)cc1
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C21H24N2O4.ClH/c1-13(11-14-3-5-15(27-2)6-4-14)22-12-19(25)16-7-9-18(24)21-17(16)8-10-20(26)23-21;/h3-10,13,19,22,24-25H,11-12H2,1-2H3,(H,23,26);1H/t13-,19+;/m1./s1
Formula Weight 404.1503
Percent Purity >95%
Chemical Name or Material Carmoterol hydrochloride
Grade HPLC