Learn More
Carl Zeiss™ Stemi 2000 Microscope Body
Description
- Apochromatic Greenough system with stereoscopic angle, 11°
- Manually operable zoom 7.7:1 (0.65x ... 5.0x) on both sides
- Switchable click stops at 0.65x, 0.8x, 1x, 1.25x, 1.6x, 2x, 2.5x, 3.2x, 4x, 4.5x
- Free working distance (without dust protection cap): 92mm
- 35° viewing angle with 55-75mm adjustable interocular distance
- 30mm eyepiece mount with maximum field number 23mm
- 76mm mount diameter
- Includes M50 dust protection cap with M49 x 0.75 for photo filter
Specifications
Specifications
| Type | Stemi™ 2000 Microscope Body |
| Includes | M50 dust protection cap with M49 x 0.75 for photo filter |