Learn More
Carl Zeiss™ Microscope Dust Cover
Specifications
Specifications
| Type | Dust Cover K |
| For Use With (Equipment) | Models: standards, Axiolab, stereomicroscopes without cameras |
| Height (Metric) | 40 cm |
| Length (Metric) | 30 cm |
| Width (Metric) | 19 cm |