Learn More
Carl Zeiss™ Immersol™ Immersion Oil
Click to view available options
Type:
Immersol 518 F
Immersol 518 N
Certifications/Compliance:
ISO 8036-1
ISO 8036-1/2
Quantity:
20 mL
100 mL
250 mL
500 mL
Packaging Type:
Bottle
Oiler
Unit Size:
100mL
20mL
250mL
500mL
Each
Description
Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
- Bubble-free immersion oil
- Suitable for oil immersion objectives from Carl Zeiss
Specifications
Specifications
| Type | Immersol 518 N |
| Certifications/Compliance | ISO 8036-1 |
| Packaging Type | Bottle |
| Quantity | 250 mL |
| Refractive Index | 1.518 |
Safety and Handling
Product Identifier
- Immersol 518 N
- Warning
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Adipic acid-di(8-methyl-tricyclo(5.2. 1.0.2.6.) decane) ester , Bis (isopropyl)naphthalene