Learn More
Carl Zeiss™ Immersol™ Immersion Oil
Product Code. 10690586
Change view
Click to view available options
Type:
Immersol 518 F
Immersol 518 N
Certifications/Compliance:
ISO 8036-1
ISO 8036-1/2
Quantity:
20 mL
100 mL
250 mL
500 mL
Packaging Type:
Bottle
Oiler
Unit Size:
100mL
20mL
250mL
500mL
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Certifications/Compliance Quantity Packaging Type Unit Size
10690586 Immersol 518 F ISO 8036-1/2 100 mL Bottle 100mL
10389092 Immersol 518 N ISO 8036-1 500 mL Bottle 500mL
13405948 Immersol 518 N ISO 8036-1 250 mL Bottle 250mL
10247272 Immersol 518 N ISO 8036-1 100 mL Bottle Each
10539438 Immersol 518 F ISO 8036-1/2 20 mL Oiler 20mL
10418653 Immersol 518 N ISO 8036-1 20 mL Oiler Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10690586 Supplier Carl Zeiss™ Supplier No. 4449620000000
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
  • Immersol 518 F
  • Bubble-free immersion oil
  • Suitable for oil immersion objectives from Carl Zeiss
Specifications
Type Immersol 518 F
Certifications/Compliance ISO 8036-1/2
Packaging Type Bottle
Quantity 100 mL
Refractive Index 1.518
compliance-icons
Product Identifier
  • Immersol 518 F
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
  • MIXTURE LIST-Contain: Adipic acid-di(8-methyl-tricyclo(5.2. 1.0.2.6.) decane) ester , Benzyl benzoate