Learn More
Zeiss™ Dust Cover Set K
Click to view available options
For Use With (Equipment):
Stemi™ microscope
Unit Size:
Each
Description
Accessory for Zeiss Microscopes
Specifications
Specifications
| Type | Dust Cover |
| Color | Blue |
| Dimensions (L x W x H) | 11.8 x 7.48 x 19.68 in. (30 x 19 x 50 cm) |
| For Use With (Equipment) | Stemi™ microscope |
| Height (English) | 19.68 in. |
| Height (Metric) | 50 cm |
| Length (English) | 11.8 in. |
| Length (Metric) | 30 cm |
| Width (English) | 7.48 in. |
| Width (Metric) | 19 cm |