Learn More
Tenak™ Origami Cardboard Cryobox
Product Code. 11571795 Shop All TENAK Products
Click to view available options
Height (Metric):
50 mm
52 mm
Includes:
10 x 10 Divider
9 x 9 Divider
No divider
Unit Size:
Pack of 100
Product Code. 11571795
Brand: Tenak™ TE22820
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Save space in the laboratory with Tenak™Origami Cardboard Cryobox. The box consists of a lid and a bottom with instructions to create a stable, high quality square cryobox..
  • Spacesaving in the laboratory
  • Square measure of 134x134mm fits all storage rack systems
  • Packages of 100 each
.
Specifications
Material Cardboard
Shape Square
Type Cryo Box
Includes 10 x 10 Divider
Quantity 100 Pack
Width (Metric) 134 mm
Height (Metric) 50 mm
Weight (Metric) 0.05 kg