Learn More
Carboxyphosphamide Benzyl Ester-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Carboxyphosphamide Benzyl Ester-d4 |
| CAS | 1276302-73-8 |
| Molecular Formula | C14H17D4Cl2N2O4P |
| Quantity | 50 mg |
| Synonym | 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Phenylmethyl Ester-d4, 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Benzyl Ester-d4 |
| SMILES | [2H]C([2H])(Cl)CN(CC([2H])([2H])Cl)P(=O)(N)OCCC(=O)OCc1ccccc1 |
| Molecular Weight (g/mol) | 387.23 |
| Formula Weight | 386.0867 |