missing translation for 'onlineSavingsMsg'
Learn More

Chemetrics™ Carbon Dioxide (dissolved) Titrets™ Titration Cells

Product Code. 11731873
Click to view available options
No. of Tests:
30
Unit Size:
Pack of 30
This item is not returnable. View return policy

Product Code. 11731873

Brand: Chemetrics™ K1925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain optimal titration with this carbon dioxide titration cells. Chemetrics™ Carbon Dioxide (dissolved) — Titrets™ Titration Cells employs a caustic titrant with pH indicator method. The titrant is a sodium hydroxide titrant and phenolphthalein indicator and a neutralizer solution to correct for sulfide interference.

  • Dependable, sealed single-use titrants
  • Fast, easy procedure -- no need to count drops
  • No burettes or other equipment necessary
  • Perform a titration

Specifications

No. of Tests 30
Range 250 to 2500ppm
Test Method Caustic Titrant with pH Indicator
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • CO2 titrets, 250-2500 mg/l
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 2
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H371-May cause damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P271-Use only outdoors or in a well-ventilated area.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P233-Keep container tightly closed.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use alcohol resistant foam or fine spray/water fog to extinguish.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P405-Store locked up.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide, sodium thiosulphate pentahydrate, phenolphthalein, methanol, ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.