missing translation for 'onlineSavingsMsg'
Learn More

Carbofuran N,N-Dibutyl-3λ1-trisulfan-1-amine, TRC

Product Code. 30385391
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385391 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385391 Supplier Toronto Research Chemicals Supplier No. TRCD7808105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbofuran N,N-Dibutyl-3Lambda1-trisulfan-1-amine
CAS 78081-78-4
Molecular Formula C20 H32 N2 O3 S3
Quantity 5 mg
Synonym 2,2-Dimethyl-2,3-dihydrobenzofuran-7-yl((dibutylamino)trisulfaneyl)(methyl)carbamate, Carbosulfan Impurity D
InChI Formula InChI=1S/C20H32N2O3S3/c1-6-8-13-22(14-9-7-2)27-28-26-21(5)19(23)24-17-12-10-11-16-15-20(3,4)25-18(16)17/h10-12H,6-9,13-15H2,1-5H3
SMILES CCCCN(CCCC)SSSN(C)C(=O)Oc1cccc2CC(C)(C)Oc12
IUPAC Name (2,2-dimethyl-3H-1-benzofuran-7-yl) N-[(dibutylamino)trisulfanyl]-N-methylcarbamate
Molecular Weight (g/mol) 444.675
Formula Weight 444.158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.