Learn More
Carbimazole, 98%, Thermo Scientific™
Product Code. p-7077743 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15590574
Brand: Thermo Scientific Acros 458610050
Request Bulk or Custom Format
Chemical Identifiers
CAS 22232-54-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
InChI Key CFOYWRHIYXMDOT-UHFFFAOYSA-N
Synonym carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
PubChem CID 31072
ChEBI CHEBI:617099
IUPAC Name ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
SMILES CCOC(=O)N1C=CN(C1=S)C
Specifications
Melting Point 122.0°C to 124.0°C
Loss on Drying 0.5% max. (24 h) (P2O5)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: 2g/L ( 20°C ). Other solubilities: soluble in acetone and ethanol
Formula Weight 186.23
Percent Purity 98%
Chemical Name or Material Carbimazole
compliance-icons
Product Identifier
  • Carbimazole
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.