Learn More
Carbazole-d8,NH, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Carbazole D8 |
| CAS | 38537-24-5 |
| Molecular Formula | C12 2H8 H N |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C12H9N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8,13 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c([nH]c3c([2 H])c([2 H])c([2 H])c([2 H])c23)c1[2 H] |
| IUPAC Name | 1,2,3,4,5,6,7,8-octadeuterio-9 H-carbazole |
| Molecular Weight (g/mol) | 175.26 |
| Formula Weight | 175.1237 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |