missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Capzimin dimer

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Capzimin dimer is a specific inhibitor of proteasome isopeptidase Rpn11 (IC50 = 2.1 μM and 3.8 μM, in 293T and A549 cell lines, respectively). It exhibits 80-fold selectivity for Rpn11 over Csn5, ten-fold over AMSH and six-fold over BRCC36 (IC50 = 30 μM, 4.5 μM, and 2.3 μM, respectively). Capzimin dimer blocks proliferation of cancer cells.
Specifications
Specifications
| Chemical Name or Material | 8,8 '-Dithiobis[N-[2-(2-thiazolyl)ethyl]-3-quinolinecarboxamide] |
| CAS | 2084868-04-0 |
| Quantity | 50 mg |
| Target | Proteasome Inhibitors |
| Molecular Formula | C30H24N6O2S4 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction