missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CAPZA1 (Human) Recombinant Protein

Human CAPZA1 full-length ORF ( AAH00144, 1 a.a. - 286 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000829-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192591

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MADFDDRVSDEEKVRIAAKFITHAPPGEFNEVFNDVRLLLNNDNLLREGAAHAFAQYNMDQFTPVKIEGYEDQVLITEHGDLGNSRFLDPRNKISFKFDHLRKEASDPQPEEADGGLKSWRESCDSALRAYVKDHYSNGFCTVYAKTIDGQQTIIACIESHQFQPKNFWNGRWRSEWKFTITPPTAQVVGVLKIQVHYYEDGNVQLVSHKDVQDSLTVSNEAQTAKEFIKIIENAENEYQTAISENYQTMSDTTFKALRRQLPVTRTKIDWNKILSYKIGKEMQNA
Specifications

Specifications

AAH00144
capping protein (actin filament) muscle Z-line, alpha 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAPZA1
GST
829
Wheat germ expression system
10 μg
CAPPA1, CAPZ, CAZ1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers