missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-CD81 Conjugate

Product Code. 30485100 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 30485100

Brand: Thermo Scientific™ 7103982100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Biotin Anti-CD81 Conjugate consists of a 12–15 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to the tetraspanin marker CD81.

CaptureSelect Biotin Anti-CD81 Conjugate consists of a 12–15 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to the tetraspanin marker CD81. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

This conjugate allows you to detect, quantitate, and characterize CD81-expressing extracellular vesicles (EVs) like exosomes. Together with CaptureSelect HRP Anti-CD81 Conjugate (Cat. No. 7303982100), a sandwich ELISA can be set up to detect and quantitate CD81 expressing EVs.

Applications for CaptureSelect Biotin Anti-CD81 Conjugate include sandwich ELISA, Gyrolab™-based immunoassays, immunoprecipitation, and sample prep purification using streptavidin-coupled Dynabeads or MSIA Streptavidin D.A.R.T.S and label-free detection platforms such as those based on surface plasmon resonance (SPR) and bio-layer interferometry (BLI). For more information see our Biotin Application Guide.

For detection of a broad variety of EVs and exosomes that originate from different cell types and secretion pathways, such as those of endosomal (CD63 enriched, CD9+, or CD81+) or ectosomal (CD63 low, CD9+ and CD81+) origin, we also offer CaptureSelect Biotin and HRP anti-CD63 conjugates and CaptureSelect Biotin and HRP anti-CD9 conjugates.

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Concentration 1 mg/mL
Formulation Phosphate-buffered saline (pH 7.4)
Content And Storage Store in freezer (–5 to –30°C).
Shipping Condition Dry Ice
Reactivity Anti-tetraspanin CD81
Form Frozen Liquid
Product Line CaptureSelect
Quantity 100 μg
Target Tetraspanin CD81
Final Product Type Biotin Conjugate
Label or Dye Biotin
Ligand Type Recombinant single-domain antibody fragment (VHH ligand)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.