Learn More
Novus Biologicals™ CaptSure DIY ELISA Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
5 x 96 Tests
Unit Size:
1 set
Description
The CaptSure DIY ELISA kit has a ready-to-go CaptSure assay plate, pre-coated with our CaptSure antibody, which is specific for the CaptSure peptide. The capture antibody is quickly and easily conjugated with the Lightning-Link CaptSure peptide supplied within the kit. Once added to the plate, the interaction between the CaptSure antibody and the CaptSure peptide immobilizes the immune-complex to the plate.
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 5 x 96 Tests |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |