Learn More
Novus Biologicals™ CaptSure DIY ELISA Kit
Product Code. 18044254 Shop All Bio Techne Products
Click to view available options
Quantity:
5 x 96 Tests
Unit Size:
1 set
Product Code. 18044254
Brand: Novus Biologicals™ 6300020
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CaptSure DIY ELISA is the latest cutting-edge ELISA kit with a built-in system for antibody labeling, enabling the rapid and easy development of in-house ELISAs using any antibody pair, while eliminating the need to perform plate coating procedures.
The CaptSure DIY ELISA kit has a ready-to-go CaptSure assay plate, pre-coated with our CaptSure antibody, which is specific for the CaptSure peptide. The capture antibody is quickly and easily conjugated with the Lightning-Link CaptSure peptide supplied within the kit. Once added to the plate, the interaction between the CaptSure antibody and the CaptSure peptide immobilizes the immune-complex to the plate.
TRUSTED_SUSTAINABILITY
Specifications
Product Type ELISA Kit
Quantity 5 x 96 Tests
Storage Requirements Storage of components varies. See protocol for specific instructions.