Learn More
Porcelaines Avignon™ Haldenwanger™ Porcelain Capsule Flat Bottom with Spout
Product Code. 11748433
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 6
Product Code. 11748433
Brand: Porcelaines Avignon™ 10107
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use this type of Porcelaines Avignon™ Porcelain Capsule lFlat Bottom with Spout in laboratories spanning from educational institutions to scientific development and industrial research..
  • Flat bottom
  • Porcelain crucible
  • White
  • Straight sides
Specifications
Description With Flat bottom spout
Bottom Shape Flat
Capacity (Metric) 100 mL
Material Porcelain
Shape Round
Type Evaporating Capsule
Height (Metric) 33 mm
Diameter (Metric) 84 mm