Learn More
BRAND™ Caps for Transferpettor™
Shop All Brand Products
Click to view available options
Volume (Metric):
5 μL
20 μL
25 μL
Unit Size:
Pack of 100
Description
- For use with Transferpettor™
- DE-M marking
- Glass construction
- Includes color code
Specifications
Specifications
| Certifications/Compliance | DE-M certified |
| Color | White |
| Material | Glass |
| Volume (Metric) | 5 μL |
| Quantity | 100 Pack |