Learn More
CAPS, MP Biomedicals
Product Code. 11453390 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25g
100g
250g
1kg
Unit Size:
100g
1kg
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453390 1kg 1kg
11423390 25g 25g
11433390 100g 100g
11443390 250g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11453390 Supplier MP Biomedicals Supplier No. 0210143591
  • ONLINE EXCLUSIVE PRICE
    625.00€
    595.35€ / 1kg
    Save 29.65€
    4% Off
 Restricted Item
Estimated Shipment: 09-11-2026