Learn More
Capping Reagent,Solstice
BIOPHARMA
Product Code. 16845421
Change view
Click to view available options
4X2.5L GLASS BOTTLE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16845421 4X2.5L GLASS BOTTLE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16845421 Supplier Burdick & Jackson BioSyn Supplier No. BR6514X2.5L
  • ONLINE EXCLUSIVE PRICE
    5280.00€
    2184.82€ / Pack of 4
    Save 3095.18€
    58% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 90/10 THF/NMI (v/v)
Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis
  • Solstice BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
CAS 616-47-7, 109-99-9
CAS Max % 10.00%, 90.00%
Quantity 4 x 2.5 L
Flash Point -15°C
Packaging Glass Bottle
Physical Form Liquid, clear
Purity Grade Notes Capping reagents
Boiling Point 66°C.The physical data is that of the main component.