Learn More
Greiner Bio-One CapMat, for use with 96-well MASTERBLOCK™ Microplates
Description
- For use with 96-well MASTERBLOCK Microplates
Specifications
Specifications
| Color | Translucent |
| Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens; resistant to DMSO; non-cytotoxic |
| Material | EVA |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| No. of Wells | 96 |
| No. per Case | 50 |
| No. per Pack | 1 |
| Pre-slit | No |
| Pierceable | No |
| Show More |