Learn More
Thermo Scientific™ Thermo Scientific™ Specimen Collection Beakers
GREENER_CHOICE
Product Code. 15628558
Change view
Click to view available options
Capacity (Metric):
103.5 mL
236 mL
Capacity (English):
3.5 oz.
8 oz.
Includes:
Pour Spout
Temperature Strip
Unit Size:
Pack of 350
Pack of 500
Product Code. Capacity (Metric) Capacity (English) Includes unitSize
15628558 103.5 mL 3.5 oz. Pour Spout Pack of 350
15638558 236 mL 8 oz. Temperature Strip Pack of 500
15648558 103.5 mL 3.5 oz. Temperature Strip Pack of 350
3 options
Product Code. 15628558
Brand: Thermo Scientific™ 10121
Thermo Scientific Collection Cups are used for for traditional and on-site testing.
Thermo Scientific™ Collection Beakers are offered in two volumes, 3 oz. and 8 oz., to support a variety of sample collection needs. The beakers feature pour spouts for easy sample transfer.
Features:
  • Pour spout for easy transfer of sample into smaller containers
  • With or without temperature indicator strips
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specimen Collection Cup
Material Polypropylene
Capacity (English) 3.5 oz.
Capacity (Metric) 103.5 mL
Description Collection Cup with pour spout; 3.5 oz. (103.5 mL)
Includes Pour Spout
No. per Case 350/Cs.