Learn More
Hirschmann™ Capillary Tubes, Both Ends Open
Product Code. 13210490
Click to view available options
Length (Metric):
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Outer:
1 mm
1.35 mm
1.55 mm
2.35 mm
Unit Size:
Pack of 1000
Pack of 250
Product Code. 13210490
missing translation for 'mfr': Hirschmann™ 9201590
Only null left
Add to basket
Add to basket
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ Capillary Tubes, Both Ends Open can be utilized for a wide range of scientific and medical applications.
  • Both ends open
  • Available in a wide range of diameters/lengths
Specifications
Diameter (Metric) Inner 1.15 mm
Diameter (Metric) Outer 1.55 mm
Tube Style Both ends open
Length (Metric) 90 mm
Type Capillary Tube