Learn More
SI Analytics™ Cannon-Fenske Reverse Flow Viscometers
Reverse flow viscometers are calibrated for manual measurement
Brand:  SI Analytics™ 285403018
Additional Details : Weight : 0.20000kg
Product Code. 10263881
  • 354.00€ / Each
Estimated Shipment: 03-06-2024
Description
  • Calibrated for manual measurement
  • Cannon-Fenske reverse flow
  • Type No. 511 03
  • Capillary No. 50
  • Constant K = 0.004
Specifications
Specifications
Cannon-Fenske Reverse Flow Viscometer