Learn More
Paragon Scientific™ Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids (UKAS ISO 17025 certified)
Product Code. 15969969
Click to view available options
Viscosity Range:
0.4 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
10,000 to 50,000 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
20,000 to 100,000 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
50 to 100 cSt
500 to 2500 cSt
7 to 35 cSt
Unit Size:
Each
Product Code. 15969969
Brand: Paragon Scientific™ CFO05UKC
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For the determination of kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
  • Supplied with full UKAS ISO 17025 calibration
Specifications
Viscosity Range 7 to 35 cSt
Calibrated Yes
Approximate Constant 0.035
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D445, ISO 3104 and IP 71 Section 1
Includes UKAS ISO 17025 calibration
Size 150
8 week lead-time