Learn More
Paragon Scientific™ Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids (UKAS ISO 17025 certified)
Click to view available options
Viscosity Range:
0.4 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
10,000 to 50,000 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
20,000 to 100,000 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
50 to 100 cSt
500 to 2500 cSt
7 to 35 cSt
Unit Size:
Each
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
- Supplied with full UKAS ISO 17025 calibration
Specifications
Specifications
| Viscosity Range | 3 to 15 cSt |
| Calibrated | Yes |
| Approximate Constant | 0.015 |
| ASTM Numbers | D 445, D 446 |
| Certifications/Compliance | ASTM D445, ISO 3104 and IP 71 Section 1 |
| Includes | UKAS ISO 17025 calibration |
| Size | 100 |
8 week lead-time