Learn More
Canertinib dihydrochloride, Tocris Bioscience™
Potent EGFR kinase inhibitor
Brand: Tocris Bioscience 5916/5
This item is not returnable.
View return policy
Description
Canertinib dihydrochloride is a potent EGFR kinase inhibitor (IC50 = 7.4 nM). Blocks autophosphorylation of erbB2. Inhibits growth of A431 tumor xenografts in mice. Cell permeable.Specifications
| N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-[3-(4-morpholinyl)propoxy]-6-quinazolinyl]-2-propenamide dihydrochloride | |
| 289499-45-2 | |
| >98% | |
| 5 mg |