Learn More
Canada Balsam, Technical Grade, TRC
Specifications
Specifications
| Chemical Name or Material | Canada Balsam, Technical Grade |
| CAS | 8007-47-4 |
| Quantity | 25 g |
| Synonym | Canada Balsams, Canada Turpentine |