Learn More
Thermo Scientific™ Campylobacter Selective Agar (Butzler)
Description
Improve isolation of Campylobacter species from fecal specimens with Thermo Scientific™ Campylobacter Selective Agar (Butzler), recommended for the isolation of Campylobacter jejuni and Campylobacter coli from clinical specimens. Supplements in the media enhance inhibition of background flora improving recovery of Campylobacter spp. which grow as flat gray colonies.
Campylobacter Selective Agar (Butzler) is supplemented with a range of antimicrobials to inhibit background flora including bacitracin, amphotericin, and novobiocin.
- Laked sheep blood supplies X factor (hemin) and other necessary growth factors
- Cefoperazone, trimethoprim, vancomycin, and polymyxin B in the medium inhibit Enterobacteriaceae, staphylococci and yeast
- Cefoperazone enhances suppression of Pseudomonas spp., thereby improving recovery of Campylobacter spp. from fecal specimens
- Ready-to-use convenience of prepared media
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 10°C |
| Description | Campylobacter Selective Agar (Butzler) |
| Format | 90mm Monoplate |
| Product Type | Agar Plate |
| Fill Weight | 19.5g ± 1g |
| Packaging Type | Wrapped in single cellulose-based film wrap |
| Quantity | 10/Pk. |