missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX cAMP Hunter™ eXpress PPYR1 (NPY4R) CHO-K1 GPCR Assay
Description
The cAMP Hunter™ PPYR1 (NPY4R) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. pancreatic polypeptide, peptide YY) based activation of PPYR1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype PPYR1 , a naturally Gi/Go coupled GPCR. Upon activation of PPYR1 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | CHO-K1 |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | Pancreatic polypeptide |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | PPYR1 (NPY4R) |
| Cell Line | CHO-K1 |
| Target | PPYR1 (NPY4R) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?