missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX cAMP Hunter™ CHO-K1 PTHR1 Gs Cell Line

Product Code. 17394619
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17394619 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17394619 Supplier DiscoverX Supplier No. 950118C2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The cAMP Hunter™ CHO-K1 PTHR1 Gs Cell Line measures PTHR1 (GPCR) activity via increased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.

The cAMP Hunter PTHR1 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. parathyroid hormone) based activation of PTHR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype PTHR1, a naturally Gs coupled GPCR. Upon activation of PTHR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Intracellular cAMP level
Cell Line CHO-K1
For Use With (Application) Cell Lines
Primary Agonist PTH(1-34)
Quantity 2 Vials
Target Entry PTHR1
Accession Number AY449732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.