missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX cAMP Hunter™ CHO-K1 MC3R Gs Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The cAMP Hunter MC3R Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. α-melanocyte stimulating hormone) based activation of MC3R GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype MC3R , a naturally Gs coupled GPCR. Upon activation of MC3R , the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Intracellular cAMP level |
| Cell Line | CHO-K1 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | α-Melanocyte stimulating hormone |
| Quantity | 2 Vials |
| Target Entry | MC3R |
| Accession Number | AY227893 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction