missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX cAMP Hunter™ CHO-K1 CNR1 Gi Cell Line

Product Code. 17374509
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17374509 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17374509 Supplier DiscoverX Supplier No. 950071C2
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The cAMP Hunter™ CHO-K1 CNR1 Gi Cell Line measures CNR1 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.

The cAMP Hunter CNR1 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CP 55,940) based activation of CNR1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype CNR1 , a naturally Gi/Go coupled GPCR. Upon activation of CNR1 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Intracellular cAMP level
Cell Line CHO-K1
For Use With (Application) Cell Lines
Primary Agonist CP 55,940
Quantity 2 Vials
Target Entry CNR1
Accession Number AY225225
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.