missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX cAMP Hunter™ CHO-K1 CHRM2 Gs Cell Line

Product Code. 17364529
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17364529

Brand: DiscoverX 950078C2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The cAMP Hunter™ CHO-K1 CHRM2 Gs Cell Line measures CHRM2 (GPCR) activity via increased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.

The cAMP Hunter CHRM2 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. oxotremorine) based activation of CHRM2 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype CHRM2 , a naturally Gs coupled GPCR. Upon activation of CHRM2 , the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Intracellular cAMP level
Cell Line CHO-K1
For Use With (Application) Cell Lines
Primary Agonist Oxotremorine-M
Quantity 2 Vials
Target Entry CHRM2
Accession Number AF498916
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.