Learn More
Carl Zeiss™ Camera Adapters
Product Code. 10397183
Click to view available options
Description:
60N-C 0.75 in.
60N-C 1.0 in.
P95-C 0.50 in.
P95-C 0.75 in.
SLR
T2-T2 DSLR
Magnification Power:
0.5x
0.63x
0.65x
1.0X
1.6X
2.5X
Unit Size:
1 unit
Each
Product Code. 10397183
Brand: Carl Zeiss™ 4155001811000
This item requires a price update prior to being ordered.
Request a Price for this item here.
C-mount camera adapter for Primovert
  • Requires Primovert
  • With a phototube C mount camera adapter
  • For Primovert
Specifications
Type Camera Adapter
Description P95-C 0.50 in.
For Use With (Equipment) Primo
Magnification Power 0.5x
Quantity 1 Ea.
Size 0.50 in.