Learn More
Carl Zeiss™ Camera Adapters
Product Code. 10337143
Change view
Click to view available options
Description:
60N-C 0.75 in.
60N-C 1.0 in.
P95-C 0.50 in.
P95-C 0.75 in.
SLR
T2-T2 DSLR
Magnification Power:
0.5x
0.63x
0.65x
1.0X
1.6X
2.5X
Unit Size:
1 unit
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Magnification Power unitSize
10337143 P95-C 0.75 in. 0.65x Each
11875873 SLR 2.5X Each
11856432 T2-T2 DSLR 1.6X Each
11895440 60N-C 0.75 in. 0.5x Each
10397183 P95-C 0.50 in. 0.5x Each
10875741 60N-C 0.75 in. 0.63x Each
15150705 60N-C 1.0 in. 1.0X 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10337143 Supplier Carl Zeiss™ Supplier No. 4155001810000
This item requires a price update prior to being ordered.
Request a Price for this item here.
C-mount camera adapter for Primovert
  • Requires Primovert
  • With a phototube C mount camera adapter
  • For Primovert
Specifications
Type Camera Adapter
Description P95-C 0.75 in.
For Use With (Equipment) Primo
Magnification Power 0.65x
Quantity 1 Ea.
Size 0.75 in.