Learn More
Calindol Amide, TRC
Product Code. 30348159
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348159 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30348159 Supplier Toronto Research Chemicals Supplier No. TRCC14865010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Calindol Amide
CAS 374933-28-5
Molecular Formula C21 H18 N2 O
Quantity 10 mg
Synonym (R)-N-[1-(1-Naphthyl)ethyl]indole-2-carboxamide, N-[(1R)-1-(1-Naphthalenyl)ethyl]-1H-Indole-2-carboxamide
InChI Formula InChI=1S/C21H18N2O/c1-14(17-11-6-9-15-7-2-4-10-18(15)17)22-21(24)20-13-16-8-3-5-12-19(16)23-20/h2-14,23H,1H3,(H,22,24)/t14-/m1/s1
SMILES C[C@@H](NC(=O)c1cc2ccccc2[nH]1)c3cccc4ccccc34
IUPAC Name N-[(1R)-1-naphthalen-1-ylethyl]-1H-indole-2-carboxamide
Molecular Weight (g/mol) 314.38
Formula Weight 314.1419