Learn More
Calindol Amide, TRC
Specifications
Specifications
| Chemical Name or Material | Calindol Amide |
| CAS | 374933-28-5 |
| Molecular Formula | C21 H18 N2 O |
| Quantity | 50 mg |
| Synonym | (R)-N-[1-(1-Naphthyl)ethyl]indole-2-carboxamide, N-[(1R)-1-(1-Naphthalenyl)ethyl]-1H-Indole-2-carboxamide |
| InChI Formula | InChI=1S/C21H18N2O/c1-14(17-11-6-9-15-7-2-4-10-18(15)17)22-21(24)20-13-16-8-3-5-12-19(16)23-20/h2-14,23H,1H3,(H,22,24)/t14-/m1/s1 |
| SMILES | C[C@@H](NC(=O)c1cc2ccccc2[nH]1)c3cccc4ccccc34 |
| IUPAC Name | N-[(1R)-1-naphthalen-1-ylethyl]-1H-indole-2-carboxamide |
| Molecular Weight (g/mol) | 314.38 |
| Formula Weight | 314.1419 |