Learn More
BINDER™ Calibration Service 1 P Pressure with Certificate for Series VD, VDL
Product Code. 17383703 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers VD/VDL Series
Unit Size:
Each
Product Code. 17383703
Brand: BINDER™ DL300410
Only null left
Add to basket
Add to basket
BINDER™ Calibration Service 1 P Pressure Service for use with vacuum drying chambers VD and VDL.
  • Calibration service includes certificate pressure
  • Calibration of the pressure display to a specified value
  • Including certificate
Specifications
Description Calibration digital pressure display - including certificate, measurement at specified pressure, range 20 - 900 Mbar
For Use With (Equipment) BINDER Chambers VD/VDL Series
Type Calibration Certificate
Display Digital
Pressure Range 20 to 900 Mbar