Learn More
BINDER™ Calibration Service 1 P Pressure with Certificate, Extension for Series VD, VDL
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers VD/VDL Series
Unit Size:
Each
Description
- Calibration service including certificate, measurement at specified pressure
- Extension of the calibration of the pressure display by another pressure value
- Including certificate
Specifications
Specifications
| Description | Calibration digital pressure display - including certificate, measurement at specified pressure, range 20 - 900 Mbar |
| For Use With (Equipment) | BINDER Chambers VD/VDL Series |
| Type | Calibration Certificate Expansion |
| Display | Digital |
| Pressure Range | 20 to 900 Mbar |