Learn More
WTW™ Calibration and Maintenance Set
Click to view available options
Type:
Standard Calibration and Maintenance Set
Technical Calibration and Maintenace Set
Unit Size:
1 set
Each
Description
- Adapted to different electrode types
- Cost-effective combinations
- Calibration and maintenance in one assortment
Specifications
Specifications
| Type | Technical Calibration and Maintenace Set |
| Includes | pH-3 Technical buffer solution 250mL (pH 4.01, pH 7, pH 10.01) trace, 2 bottles of 250mL of KCl 3mol/L bottles |
| For Use With (Equipment) | Gel electrodes |