missing translation for 'onlineSavingsMsg'
Learn More

Xylem AG Refractometer Calibration Fluid 20 x 5 mL (UKAS/NIST)

Product Code. 30484465
Click to view available options
Refractive Index:
1.33659
1.34026
1.34401
1.34782
1.34968
1.35093
1.35171
1.35568
1.39986
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 30484465

Brand: Xylem 90603

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Calibration fluid for calibration of refractometers.

  • Calibration Fluid
  • For Refractometers
TRUSTED_SUSTAINABILITY

Specifications

Type Calibration Fluid
Refractive Index 1.34401
For Use With (Equipment) Refractometer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.