Learn More
BINDER™ Calibration Certificate, Temperature and Humidity
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series MKF
BINDER Chambers Series MKFT
Calibration Points:
1 Measuring Point
27 Measuring Points
Unit Size:
Each
Description
- One humidity point of measurement
- Including certificate
- Humidity measurement point in center of chamber
- Temperature (according to DIN12880) and humidity measurement at 25°C / 60% RH or at specified values
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series MKF |
| Description | Calibration certificate, temperature and humidity - Measurement in center of chamber at 25°C / 60% RH or at specified test values |
| Calibration Points | 1 Measuring Point |
| Product Type | Calibration Certificate (Temperature and Humidity) |
| For Use With (Application) | Temperature and Humidity Measurement |