Learn More
BINDER™ Calibration Certificate for BINDER™ KB ECO Series
Product Code. 17512875 Shop All BINDER Products
Click to view available options
Calibration Type:
1 P Expanded Temperature
1 P Temperature
18 P Temperature
27 P Temperature
9 P Temperature
Unit Size:
Each
Product Code. 17512875
Brand: BINDER™ 80122107
The Calibration Certificate for BINDER KB ECO Series is an option for KB ECO Series Refrigerated Incubators with environmentally friendly Thermoelectric Cooling.
  • Calibration certificate
  • For temperature
Specifications
For Use With (Equipment) BINDER Chambers Series KB ECO
Description Calibration certificate, expanded - for temperature; for extending the measurement in center of chamber to include another test temperature
Duration 60 to 120 min.
Calibration Type 1 P Expanded Temperature
Product Type Calibration Certificate