Learn More
BINDER™ Calibration Certificate for BINDER™ KB ECO Series
Shop All BINDER Products
Click to view available options
Calibration Type:
1 P Expanded Temperature
1 P Temperature
18 P Temperature
27 P Temperature
9 P Temperature
Unit Size:
Each
Description
- Calibration certificate
- For temperature
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series KB ECO |
| Description | Calibration certificate, expanded - for temperature; for extending the measurement in center of chamber to include another test temperature |
| Duration | 60 to 120 min. |
| Calibration Type | 1 P Expanded Temperature |
| Product Type | Calibration Certificate |